missing translation for 'onlineSavingsMsg'
Learn More

Methyl (S)-(+)-3-hydroxybutyrate, 98%

Product Code. 11374376
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11374376 5 g 5g
11364376 1 g 1g
11384376 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11374376 Supplier Thermo Scientific Alfa Aesar Supplier No. L19996.06
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 53562-86-0 | C5H10O3 | 118.13 g/mol

Methyl (S)-(+)-3-hydroxybutyrate serves as an optically active starting material used in organic synthesis. It is also used in the preparation of (-)-methyl elenolate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl (S)-(+)-3-hydroxybutyrate serves as an optically active starting material used in organic synthesis. It is also used in the preparation of (-)-methyl elenolate.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Incompatible with oxidizing agents, bases and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53562-86-0
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00064461
InChI Key LDLDJEAVRNAEBW-UHFFFAOYNA-N
Synonym methyl s-+-3-hydroxybutyrate, s-methyl 3-hydroxybutanoate, methyl s-3-hydroxybutyrate, s-3-hydroxybutanoic acid methyl ester, unii-g10281a5ah, methyl 3s-3-hydroxybutanoate, butanoic acid, 3-hydroxy-, methyl ester, 3s, methyl-s-3-hydroxybutyrate, s-+-3-hydroxy-n-butyric acid methyl ester, s-3-hydroxybutyric acid methyl ester
PubChem CID 6950307
IUPAC Name methyl (3S)-3-hydroxybutanoate
SMILES COC(=O)CC(C)O

Specifications

Density 1.055
Boiling Point 56°C to 58°C (11 mmHg)
Flash Point 73°C (163°F)
Refractive Index 1.422
Quantity 5 g
Beilstein 1720583
Solubility Information Miscible with alcohol. Immiscible with water.
Optical Rotation +48.5° (c=1.3 in Chloroform)
Formula Weight 118.13
Percent Purity 98%
Chemical Name or Material Methyl (S)-(+)-3-hydroxybutyrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.