missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Undecanoate-d21, CDN
Click to view available options
Quantity:
0.1 g
0.5 g
Unit Size:
0.1g
0.5g
Specifications
Specifications
| Chemical Name or Material | Methyl Undecanoate-D21 |
| CAS | 1219804-96-2 |
| Molecular Formula | C12 D21 H3 O2 |
| Quantity | 0.1 g |
| Synonym | Methyl Undecanoate-d21, Undecanoic-d21 acid methyl ester, Undecanoic acid methyl ester-d21, Methyl Undecanoate-D21 |
| InChI Formula | InChI=1 S/C12H24O2/c1-3-4-5-6-7-8-9-10-11-12(13)14-2/h3-11H2,1-2H3/i1D3,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)OC |
| IUPAC Name | methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-henicosadeuterioundecanoate |
| Molecular Weight (g/mol) | 221.4472 |
| Formula Weight | 221.3094 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction