missing translation for 'onlineSavingsMsg'
Learn More

Methyl Violet, Pure, C.I.42555, Fisher Chemical

Product Code. 10142150 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10142150 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10142150 Supplier Fisher Chemical Supplier No. M/5350/48

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 215°C
CAS 548-62-9
Molecular Formula C25H30ClN3
MDL Number 11750
InChI Key ZXJXZNDDNMQXFV-UHFFFAOYSA-M
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
Molecular Weight (g/mol) 407.986
PubChem CID 11057
ChEBI CHEBI:41688
Formula Weight 407.97g/mol
Packaging Glass Jar
Color Green
Quantity 100 g
Physical Form Solid
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methyl violet
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Carcinogenicity Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P273-Avoid release to the environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.