missing translation for 'onlineSavingsMsg'
Learn More

Methylamine (2.0M in methanol), TRC

Product Code. 30477818
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477818 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477818 Supplier Toronto Research Chemicals Supplier No. TRCM28556010ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methanamine
CAS 74-89-5
Molecular Formula C H5 N
Quantity 10 mL
Synonym Methanamine, Methylamine (8CI), Aminomethane, Carbinamine, MMA, Monomethylamine, R 630
InChI Formula InChI=1S/CH5N/c1-2/h2H2,1H3
SMILES NC
IUPAC Name methanamine
Molecular Weight (g/mol) 31.06
Formula Weight 31.0422
Concentration or Composition (by Analyte or Components) 2 M
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.