missing translation for 'onlineSavingsMsg'
Learn More

[(Methylamino)methyl] Phosphonic Acid, TRC

Product Code. 17930825
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17930825 50 mg 50mg
17964795 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17930825 Supplier Toronto Research Chemicals Supplier No. TRCM28695050MG
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35404-71-8
Molecular Formula C2 H8 N O3 P
Molecular Weight (g/mol) 125.064
Synonym P-[(Methylamino)methyl]phosphonic Acid, [(Methylamino)methyl]phosphonic Acid, NSC 244826
IUPAC Name methylaminomethylphosphonic acid
SMILES CNCP(=O)(O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C2H8NO3P/c1-3-2-7(4,5)6/h3H,2H2,1H3,(H2,4,5,6)
Formula Weight 125.0242
Chemical Name or Material [(Methylamino)methyl] Phosphonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.