missing translation for 'onlineSavingsMsg'
Learn More

Methylcyclohexane, TRC

Product Code. 30466780
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30466780 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30466780 Supplier Toronto Research Chemicals Supplier No. TRCM338318500G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methylcyclohexane
CAS 108-87-2
Molecular Formula C7 H14
Quantity 500 g
Synonym Cyclohexane, methyl-, Methylcyclohexane, 1-Methylcyclohexane, Cyclohexylmethane, Hexahydrotoluene, MCHX, NSC 9391, Swaclean MCH, Toluene hexahydride
InChI Formula InChI=1S/C7H14/c1-7-5-3-2-4-6-7/h7H,2-6H2,1H3
SMILES CC1CCCCC1
IUPAC Name methylcyclohexane
Molecular Weight (g/mol) 98.19
Formula Weight 98.1096
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.