missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylcyclopentane-d12, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Methylcyclopentane-D12 |
| CAS | 144120-51-4 |
| Molecular Formula | C6 D12 |
| Quantity | 1 g |
| Synonym | Cyclopentane-d9, methyl-d3- (9 CI), Methyl-d3-cyclopentane-d9, Methylcyclopentane-d12, Methylcyclopentane-D12 |
| InChI Formula | InChI=1 S/C6H12/c1-6-4-2-3-5-6/h6H,2-5H2,1H3/i1D3,2D2,3D2,4D2,5D2,6 D |
| SMILES | [2 H]C([2 H])([2 H])C1([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C1([2 H])[2 H] |
| IUPAC Name | 1,1,2,2,3,3,4,4,5-nonadeuterio-5-(trideuteriomethyl)cyclopentane |
| Molecular Weight (g/mol) | 96.2334 |
| Formula Weight | 96.1692 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction