missing translation for 'onlineSavingsMsg'
Learn More

Methyldiphenylphosphine oxide, 98+%

Product Code. 11484320
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484320 25 g 25g
11474320 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484320 Supplier Thermo Scientific Alfa Aesar Supplier No. A11484.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 2129-89-7 | C13H13OP | 216.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2129-89-7
Molecular Formula C13H13OP
Molecular Weight (g/mol) 216.22
MDL Number MFCD00002081
InChI Key PEGCITODQASXKH-UHFFFAOYSA-N
Synonym methyldiphenylphosphine oxide, methyl diphenyl phosphine oxide, diphenylmethylphosphine oxide, phosphine oxide, methyldiphenyl, phosphine oxide, diphenylmethyl, methyl-phenylphosphoryl benzene, methyl phenyl phosphoryl benzene, methyl phenyl phosphoroso benzene, methyldiphenylphosphineoxide
PubChem CID 75041
IUPAC Name [methyl(phenyl)phosphoryl]benzene
SMILES CP(=O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 111°C to 114°C
Quantity 25 g
Beilstein 957291
Formula Weight 216.22
Percent Purity ≥98%
Chemical Name or Material Methyldiphenylphosphine oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.