missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylhydroquinone, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Methylhydroquinone |
| CAS | 95-71-6 |
| Molecular Formula | C7H8O2 |
| Quantity | 10 g |
| Synonym | 2,5-Dihydroxytoluene, 2,5-Toluenediol, 2-Methyl-1,4-benzenediol, 2-Methyl-1,4-dihydroxybenzene, 2-Methyl-1,4-hydroquinone, 2-Methyl-p-hydroquinone, 2-Methylhydroquinone, 4-Hydroxy-2-methylphenol, M-HQ, Methyl 1,4-dihydroxybenzene, Methyl-p-hydroquinone, Methylhydroquinone, NSC 4962, THQ, Toluhydroquinone, Tolylhydroquinone, m-Methyl-hydroquinone, o-Methylhydroquinone, p-Toluhydroquinol, p-Toluhydroquinone, p-Toluquinol, 1,4-Dihydroxy-2-methylbenzene |
| InChI Formula | InChI=1S/C7H8O2/c1-5-4-6(8)2-3-7(5)9/h2-4,8-9H,1H3 |
| SMILES | Cc1cc(O)ccc1O |
| Molecular Weight (g/mol) | 124.14 |
| Formula Weight | 124.0524 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction