missing translation for 'onlineSavingsMsg'
Learn More

Methylnadic Anhydride, Serva Electrophoresis™

Product Code. 11741448
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11741448 100 mL Each
11751448 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11741448 Supplier Serva Electrophoresis Supplier No. 2945201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 25134-21-8
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
InChI Key KNRCVAANTQNTPT-UHFFFAOYNA-N
Synonym methy nadic anhydride mna, 3ar,4s,7r,7as-3a-methyl-3a,4,7,7a-tetrahydro-4,7-methano-2-benzofuran-1,3-dione, methylnadic anhydride, dsstox_cid_7844, dsstox_rid_78588, dsstox_gsid_27844, 3a-methyl-3a,4,7,7a-tetrahydro-4,7-methanoisobenzofuran-1,3-dione relative, 3abeta-methyl-3a,4,7,7abeta-tetrahydro-4beta,7beta-methanoisobenzofuran-1,3-dione

Specifications

Quantity 100 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methylnadic anhydride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 3
  • Respiratory sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.