missing translation for 'onlineSavingsMsg'
Learn More

Methylsulfonylacetonitrile, 98+%

Product Code. 11352086
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11352086 50 g 50g
11342086 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352086 Supplier Thermo Scientific Alfa Aesar Supplier No. L03280.18
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2274-42-2 | C3H5NO2S | 119.138 g/mol

(Methylsulfonyl)acetonitrile was used in the synthesis of substituted δ-pyrone ring analogues, 4,9-dihydropyrrolo[2,1-b]quinazolines containing electron withdrawing groups at the 3-position.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Methylsulfonyl)acetonitrile was used in the synthesis of substituted δ-pyrone ring analogues, 4,9-dihydropyrrolo[2,1-b]quinazolines containing electron withdrawing groups at the 3-position.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2274-42-2
Molecular Formula C3H5NO2S
Molecular Weight (g/mol) 119.138
MDL Number MFCD00014743
InChI Key FOTRKCAZUSJCQD-UHFFFAOYSA-N
Synonym methylsulfonylacetonitrile, 2-methylsulfonyl acetonitrile, methylsulfonyl acetonitrile, methanesulfonylacetonitrile, mesylacetonitrile, 2-methanesulfonylacetonitrile, cyanomethylsulfonylmethane, methanesulphonylacetonitrile, acetonitrile, methylsulfonyl, cyanomethyl methyl sulphone
PubChem CID 75283
ChEBI CHEBI:32444
IUPAC Name 2-methylsulfonylacetonitrile
SMILES CS(=O)(=O)CC#N

Specifications

Melting Point 82°C to 86°C
Quantity 50 g
Beilstein 969929
Solubility Information Slightly soluble in water.
Formula Weight 119.14
Percent Purity ≥98%
Chemical Name or Material Methylsulfonylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.