missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyltri-n-butylammonium bromide, 98%, Thermo Scientific™

Product Code. 11440298
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440298 50 g 50g
11430298 10 g 10g
11450298 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440298 Supplier Alfa Aesar™ Supplier No. B23828.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37026-88-3
Molecular Formula C13H30BrN
Molecular Weight (g/mol) 280.29
MDL Number MFCD00043201
InChI Key FQUHTZCWXDFLBQ-UHFFFAOYSA-N
Synonym tributylmethylammonium bromide, methyltri-n-butylammonium bromide, 1-butanaminium, n,n-dibutyl-n-methyl-, bromide, methyltributylammonium bromide, methyl tributyl ammonium bromide, tributyl methyl azanium bromide, acmc-20ajfa, ksc493m4d, tributylmethylammonium bro-mide
PubChem CID 10062191
IUPAC Name tributyl(methyl)azanium;bromide
SMILES [Br-].CCCCC([NH3+])(CCCC)CCCC

Specifications

Melting Point 120°C to 122°C
Quantity 50 g
Beilstein 3704268
Sensitivity Hygroscopic
Formula Weight 280.29
Percent Purity 98%
Chemical Name or Material Methyltri-n-butylammonium bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.