missing translation for 'onlineSavingsMsg'
Learn More

Methyltrioctylammonium Chloride, TRC

Product Code. 30137303
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137303 50 g 50g
30133961 1 g 1g
30006715 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137303 Supplier Toronto Research Chemicals Supplier No. TRCM33120550G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5137-55-3
Molecular Formula C25 H54 N . Cl
Molecular Weight (g/mol) 404.16
Synonym Methyltrioctylammonium Chloride, A 336, A 336 (catalyst), Aliquat 128, Capriquat, Methyltri-n-octylammonium Chloride, Methyltricaprylammonium Chloride, N-Methyl-N,N,N-trioctylammonium Chloride, TOMAC, TOMAC 75, Tri-n-octylmethylammonium Chloride, Tricaprylmethylammonium Chloride, Trioctylmonomethylammonium Chloride;
IUPAC Name methyl(trioctyl)azanium;chloride
SMILES [Cl-].CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC

Specifications

Quantity 50 g
InChI Formula InChI=1S/C25H54N.ClH/c1-5-8-11-14-17-20-23-26(4,24-21-18-15-12-9-6-2)25-22-19-16-13-10-7-3;/h5-25H2,1-4H3;1H/q+1;/p-1
Formula Weight 403.3945
Chemical Name or Material Methyltrioctylammonium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.