missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metoclopramide Hydrochloride Monohydrate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Metoclopramide Hydrochloride Monohydrate |
| CAS | 54143-57-6 |
| Analyte or Component Names | Metoclopramide Hydrochloride Monohydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H22 Cl N3 O2 . Cl H . H2 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | Metoclopramide Hydrochloride Monohydrate, 4-Amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide monohydrochloride monohydrate |
| InChI Formula | InChI=1S/C14H22ClN3O2.ClH.H2O/c1-4-18(5-2)7-6-17-14(19)10-8-11(15)12(16)9-13(10)20-3;;/h8-9H,4-7,16H2,1-3H3,(H,17,19);1H;1H2 |
| Solution Type | Neat |
| SMILES | O.Cl.CCN(CC)CCNC(=O)c1cc(Cl)c(N)cc1OC |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?