missing translation for 'onlineSavingsMsg'
Learn More

Metoclopramide hydrochloride, Thermo Scientific™

5-HT3 serotonin antagonist; D2 dopamine antagonist

Brand:  Thermo Scientific Alfa Aesar J61545.22

Quantity 100g

 View more versions of this product

Product Code. 15404139

  • 225.00€ / 100g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Metoclopramide hydrochloride is used as a digestive system drugs. It is used for anti-emetic, used for digestive dysfunction.

Solubility
It is very soluble in water, easily soluble in ethyl alcohol, slightly soluble in dichloromethane.

Notes
Hygroscopic, Refrigerator, Under Inert atmosphere. Incompatible to strong alkalis, strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

54143-57-6
354.27
KJBLQGHJOCAOJP-UHFFFAOYSA-N
CHEBI:6899
O.Cl.CCN(CC)CCNC(=O)C1=CC(Cl)=C(N)C=C1OC
C14H25Cl2N3O3
MFCD00150634
441347
4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide hydrate hydrochloride

Specifications

54143-57-6
MFCD00150634
Light sensitive
It is very soluble in water,easily soluble in ethyl alcohol,slightly soluble in dichloromethane.
O.Cl.CCN(CC)CCNC(=O)C1=CC(Cl)=C(N)C=C1OC
354.27
CHEBI:6899
Metoclopramide hydrochloride
C14H25Cl2N3O3
100g
14,6143
KJBLQGHJOCAOJP-UHFFFAOYSA-N
4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide hydrate hydrochloride
441347
336.26
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.