Learn More
Metoclopramide hydrochloride, Thermo Scientific™
5-HT3 serotonin antagonist; D2 dopamine antagonist
44.40€ - 225.00€
Chemical Identifiers
| CAS | 54143-57-6 |
|---|---|
| Molecular Formula | C14H25Cl2N3O3 |
| Molecular Weight (g/mol) | 354.27 |
| MDL Number | MFCD00150634 |
| InChI Key | KJBLQGHJOCAOJP-UHFFFAOYSA-N |
| PubChem CID | 441347 |
| ChEBI | CHEBI:6899 |
| IUPAC Name | 4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide hydrate hydrochloride |
| SMILES | O.Cl.CCN(CC)CCNC(=O)C1=CC(Cl)=C(N)C=C1OC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15484129
|
Thermo Scientific Alfa Aesar
J61545.09 |
10g |
44.40€
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15494129
|
Thermo Scientific Alfa Aesar
J61545.14 |
25g |
88.00€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15404139
|
Thermo Scientific Alfa Aesar
J61545.22 |
100g |
225.00€
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsMetoclopramide hydrochloride is used as a digestive system drugs. It is used for anti-emetic, used for digestive dysfunction.
Solubility
It is very soluble in water, easily soluble in ethyl alcohol, slightly soluble in dichloromethane.
Notes
Hygroscopic, Refrigerator, Under Inert atmosphere. Incompatible to strong alkalis, strong oxidizing agents.
Chemical Identifiers
| 54143-57-6 | |
| 354.27 | |
| KJBLQGHJOCAOJP-UHFFFAOYSA-N | |
| CHEBI:6899 | |
| O.Cl.CCN(CC)CCNC(=O)C1=CC(Cl)=C(N)C=C1OC |
| C14H25Cl2N3O3 | |
| MFCD00150634 | |
| 441347 | |
| 4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide hydrate hydrochloride |
Specifications
| 54143-57-6 | |
| MFCD00150634 | |
| 14,6143 | |
| KJBLQGHJOCAOJP-UHFFFAOYSA-N | |
| 4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide hydrate hydrochloride | |
| 441347 | |
| 336.26 |
| C14H25Cl2N3O3 | |
| Light sensitive | |
| It is very soluble in water,easily soluble in ethyl alcohol,slightly soluble in dichloromethane. | |
| O.Cl.CCN(CC)CCNC(=O)C1=CC(Cl)=C(N)C=C1OC | |
| 354.27 | |
| CHEBI:6899 | |
| Metoclopramide hydrochloride |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.