missing translation for 'onlineSavingsMsg'
Learn More

Metoprolol Succinate, TRC

Product Code. 30135001
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135001 1 g 1g
30135126 5 g 5g
30004800 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135001 Supplier Toronto Research Chemicals Supplier No. TRCM3388251G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98418-47-4
Molecular Formula 2 C15 H25 N O3 . C4 H6 O4
Molecular Weight (g/mol) 652.82
Synonym Metoprolol Succinate, Butanedioic acid compd. with 1-[4-(2-methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]-2-propanol (1:2), Butanedioic acid compd. with (±)-1-[4-(2-methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]-2-propanol (1:2), (±)-1-[4-(2-Methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]-2-propanol butanedioate (2:1) (salt), 1-[4-(2-Methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]-2-propanol butanedioate (2:1) (salt), Beloc-Zok, Betaloc CR, Betaloc Zok, Betazok, H 93/26 succinate, Metoprolol succinate, Seloken-Zok, Selozok, Toprol XL
IUPAC Name butanedioic acid;1-[4-(2-methoxyethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol
SMILES COCCc1ccc(OCC(O)CNC(C)C)cc1.COCCc2ccc(OCC(O)CNC(C)C)cc2.OC(=O)CCC(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/2C15H25NO3.C4H6O4/c2*1-12(2)16-10-14(17)11-19-15-6-4-13(5-7-15)8-9-18-3;5-3(6)1-2-4(7)8/h2*4-7,12,14,16-17H,8-11H2,1-3H3;1-2H2,(H,5,6)(H,7,8)
Formula Weight 652.3935
Chemical Name or Material Metoprolol Succinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.