missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ METRNL Monoclonal Antibody (829519)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561704
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Hormone induced following exercise or cold exposure that promotes energy expenditure. Induced either in the skeletal muscle after exercise or in adipose tissue following cold exposure and is present in the circulation. Able to stimulate energy expenditure associated with the browning of the white fat depots and improves glucose tolerance. Does not promote an increase in a thermogenic gene program via direct action on adipocytes, but acts by stimulating several immune cell subtypes to enter the adipose tissue and activate their prothermogenic actions. Stimulates an eosinophil-dependent increase in IL4 expression and promotes alternative activation of adipose tissue macrophages, which are required for the increased expression of the thermogenic and anti-inflammatory gene programs in fat. Required for some cold-induced thermogenic responses, suggesting a role in metabolic adaptations to cold temperatures.
Specifications
| METRNL | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8VE43 | |
| METRNL | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Meteorin-like/METRNL, Gln46-Glu311. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Frozen) | |
| 829519 | |
| Unconjugated | |
| METRNL | |
| 9430048M07Rik; BC019776; cometin; meteorin like, glial cell differentiation regulator; meteorin, glial cell differentiation regulator-like; meteorin-like protein; METRNL; Subfatin | |
| Rat | |
| Protein A/G | |
| RUO | |
| 210029 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction