Learn More
Metro™ Double-Deep Top-Track Stationary Intermediate Unit Kits
Product Code. 15967151
Change view
Click to view available options
Width (English):
18 in.
21 in.
24 in.
Width (English) Shelf:
18 in.
21 in.
24 in.
Weight (English):
18 lb.
20 lb.
22 lb.
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (English) Width (English) Shelf Weight (English) Unit Size
15967151 24 in. 24 in. 22 lb. Each
15947151 18 in. 18 in. 18 lb. Each
15957151 21 in. 21 in. 20 lb. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15967151 Supplier Metro™ Supplier No. LTTA24C
  • ONLINE EXCLUSIVE PRICE
    809.00€
    645.97€ / Pack of 1
    Save 163.03€
    20% Off
Estimated Shipment: 21-04-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Connects intermediate units in Double-Deep systems and to overhead track sets
  • For use with Metro Super Erecta™ and Super Adjustable™ shelving systems
  • Chrome-plated finish
  • Kit includes connecting hardware
Order Info
One kit is required for intermediate units connected end-to-end. Shelves are sold separately.
Specifications
Product Type Stationary Intermediate Unit Kit
Finish Chrome
For Use With (Equipment) Metro™ Super Erecta™, Super Adjustable™ shelving system
Includes 86 in. Posts and connecting hardware
Weight (English) 22 lb.
Weight (Metric) 9.9 kg
Width (English) Shelf 24 in.
Width (English) 24 in.
Width (Metric) Shelf 61 cm
Width (Metric) 61 cm
Material Steel