Learn More
Metro™ Lab Cart Accessory, Autoclavable Cover
Product Code. 11715699
Change view
Click to view available options
Width (English):
26 in.
Length (English):
38 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (English) Length (English) Unit Size
11715699 26 in. 38 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11715699 Supplier Metro™ Supplier No. LC2436AC
  • ONLINE EXCLUSIVE PRICE
    751.00€
    601.90€ / Pack of 1
    Save 149.10€
    19% Off
 Restricted Item
Estimated Shipment: 03-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Lab cart cover is sized to fit Metro autoclavable lab carts.
  • Rated for approximately 70 cycles per cover
Specifications
Product Type Cart Cover
Color Blue
Dimensions (L x W x H) 38 x 26 x 33.5 in. (96.5 x 66 x 85.1 cm)
For Use With (Equipment) 24 in. W Lab cart
Height (English) 33.5 in.
Height (Metric) 85.1 cm
Length (English) 38 in.
Length (Metric) 96.5 cm
Width (English) 26 in.
Width (Metric) 66 cm