Learn More
Metro™ Super Adjustable Super Erecta™ Wire Shelving Accessory, 8 in. Shelf Dividers
Product Code. 11791049
Change view
Click to view available options
Width (English):
18 in.
24 in.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (English) unitSize
11791049 18 in. Each
11701059 24 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11791049 Supplier Metro™ Supplier No. DD18C
Only null left
Add to basket
Add to basket
Tall dividers (8 in.) snap onto wire shelves to partition shelf space for storage optimization.
  • Dividers can also attach to the top or bottom of shelves.
  • Available in Chrome or Type 304 Stainless Steel.
  • Other sizes available.
Specifications
Product Type Shelf Divider for Wire Shelves
For Use With (Equipment) Metro™ Super Erecta™ shelving
Height (English) 8 in.
Height (Metric) 20.3 cm
Width (English) 18 in.
Width (Metric) 45.7 cm
Material Chrome