Learn More
Metro™ Super Erecta™ Top Track Mobile Unit Kits
Product Code. 15977301
Change view
Click to view available options
Material:
Stainless Steel
Steel
Width (English) Shelf:
18 in.
21 in.
24 in.
Finish:
Chrome
Metroseal 3 with Microban™
Metroseal Green Epoxy
None
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Width (English) Shelf Finish Unit Size
15977301 Stainless Steel 24 in. None Each
15917301 Stainless Steel 18 in. None Each
15947301 Stainless Steel 21 in. None Each
15927301 Steel 21 in. Chrome Each
15957301 Steel 24 in. Chrome Each
15907301 Steel 18 in. Metroseal 3 with Microban™ Each
15937301 Steel 21 in. Metroseal 3 with Microban™ Each
15967301 Steel 24 in. Metroseal Green Epoxy Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15977301 Supplier Metro™ Supplier No. TTM24S
  • ONLINE EXCLUSIVE PRICE
    965.00€
    771.53€ / Pack of 1
    Save 193.47€
    20% Off
Estimated Shipment: 21-04-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Includes
Posts, casters/caster channels, donut bumpers and roller bearing assemblies
High-density, easy access storage.
Top-Track storage systems increase storage capacity and ease of access with overhead guide tracks. This design allows for open aisles eliminating hard to reach spaces, an interchangeable/modular storage system and easier cleaning.
  • Mobile units move along anchored track allowing easy access to all shelves
Order Info
Shelves are ordered separately. One kit required per mobile unit.
Specifications
Product Type Mobile Unit Kit
Finish None
For Use With (Equipment) Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system
Includes Posts, casters/channel caster, donut bumpers and roller bearing assemblies, hardware, split sleeves
Weight (English) 34 lb.
Weight (Metric) 15.3 kg
Width (English) Shelf 24 in.
Width (English) 26.25 in.
Width (Metric) Shelf 61 cm
Width (Metric) 66.7 cm
Material Stainless Steel