Learn More
METTL7A Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | METTL7A |
|---|---|
| Concentration | 267 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16856934
|
Proteintech
17092-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16846934
|
Proteintech
17092-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The METTL7A antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human METTL7A. This antibody recognizes human, mouse, rat antigen. The METTL7A antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| METTL7A | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG |
| 267 μg/mL | |
| Polyclonal | |
| Rabbit | |
| 25840 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.