Learn More
Mettler Toledo™ Compact Stirrer, for Mettler Toledo™ Titrators
Product Code. 10700530 Shop All Mettler Toledo Products
Click to view available options
For Use With (Equipment):
DL15/22/28 titrators
Unit Size:
Each
Product Code. 10700530
Brand: Mettler Toledo™ 51109150
Only null left
Add to basket
Add to basket
Accessory for use with DL22 Food and Beverage Titrator and DL15 and DL18 titrators.
Accessory for use with DL22 Food and Beverage Titrator and DL15 and DL18 titrators.
TRUSTED_SUSTAINABILITY
Specifications
Type Compact Overhead Stirrer with Two Propellers
For Use With (Application) Titration
Includes 2 Propeller stirrers
For Use With (Equipment) DL15/22/28 titrators