Learn More
METTLER TOLEDO™ InLab™ Versatile pH Electrode Replaceable Ceramic Junctions
Product Code. 12669195 Shop All Mettler Toledo Products
Change view
Click to view available options
Junction:
Ceramic
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Junction unitSize
12669195 Ceramic Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12669195 Supplier METTLER TOLEDO™ Supplier No. 51340268
Only null left
Add to basket
Add to basket
Set of 10 ceramic junctions.
  • Accessory
  • For use with InLab™ Versatile and InLab™ Versatile Pro
TRUSTED_SUSTAINABILITY
Specifications
Junction Ceramic
Temperature (Metric) Operating 0°C to 60°C
Type Set of replaceable junctions
For Use With (Application) Alternative for measurements in simple samples, offer an outstanding price/performance ratio
For Use With (Equipment) InLab designations versatile and versatile Pro pH electrode
Includes 10 Ceramic junction