Learn More
Mettler Toledo™ Ion-Selective Combination Electrodes: Filling Solutions
Product Code. 12609195 Shop All Mettler Toledo Products
Click to view available options
Filling Solution Type:
0.5 mol/L MgSO4
1 mol/L KNO3
3 mol/L KCl
For Use With (Equipment):
Ammonium ISE
Bromide ISE
Calcium ISE
Chloride ISE
Fluoride ISE
Nitrate ISE
Potassium ISE
Unit Size:
20mL
Product Code. 12609195
Brand: Mettler Toledo™ 51340033
For use with ISE combination electrodes.
For use with ISE combination electrodes.
TRUSTED_SUSTAINABILITY
Specifications
Description Electrolyte for Potassium ISE, 20mL
For Use With (Equipment) Potassium ISE
Type ISE Filling Solution
Filling Solution Type 0.5 mol/L MgSO4
For Use With (Application) Determine the Potassium concentration in samples
Packaging Bottle
Quantity 20 mL
Temperature (English) Operating 32°F to 122°F
Temperature (Metric) Operating 0°C to 50°C