Learn More
METTLER TOLEDO™ SevenExcellence™ S500 pH/ISE Benchtop Meter
Product Code. 13490679 Shop All Mettler Toledo Products
Click to view available options
Product Type:
Meter with Bio Kit
Meter with Fluoride Kit
Meter with Standard Kit
pH/ISE Meter
Unit Size:
Each
Product Code. 13490679
Brand: METTLER TOLEDO™ 30046251
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
The S500 meter performs ion selective (ISE), pH, redox/ORP, and ISFET measurements. It can be expanded with additional modules to measure conductivity, resistivity, DO, salinity, TDS, and BOD. It has an intuitive touchscreen and the ability to create measurement shortcuts.
  • Ion selective (ISE), pH, redox/ORP, and ISFET meter than can be expanded with additional measurement modules to measure conductivity, resistivity, dissolved oxygen, salinity, TDS, and BOD
  • Easily create measurement and calibration shortcuts
  • 7 inch color touch screen with intuitive menu navigation
  • Compatible with a wide range of electrodes for any application
  • Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
  • Meter connects to electrodes via BNC; S7 to BNC cables available
  • Export data to a printer, Excel file, or LIMS system
  • Automation available for measurements and cleaning in high-throughput applications
  • Manage user permissions and access to ensure data integrity
  • Compatible with LabX software for advanced data management and 21CFR part 11 regulatory compliance
  • Optional comprehensive qualification package available including IQ/OQ/PQ
TRUSTED_SUSTAINABILITY
Specifications
Outputs RS232, USB-A, USB-B, LAN, Mini-LTW, Mini-DIN, BNC, Cinch/RCA
Accuracy (Concentration) ±0.5%
Accuracy (mV) ±0.1mV
Accuracy (pH) ±0.002
Calibration Points (Conductivity) 13 Pre defined and 20 user defined standard
Calibration Points (ISE) Max. 5 points, 8 pre defined and 20 user defined buffer group
Calibration Points (pH) Max. 5 points, 8 pre defined and 20 user defined buffer group
Data Points 20,000
Channels 1
Display Type TFT
Range (Conductivity) 0.001 μS/cm to 2000 mS/cm
Range (mV) -2000 to 2000 mV
Range (pH) -2 to 20
Range (Resistivity) 0.001 μS/cm to 2000 mS/cm
Range (Salinity) 0.001 μS/cm to 2000 mS/cm
Range (TDS) 0.001 μS/cm to 2000 mS/cm
Resolution (ISE) 0.001 to 1
Resolution (pH) 0.001/0.01/0.1
Temperature Range (Metric) -30°C to +130°C
Includes S500 pH/ISE meter, perfectION comb F ion selective electrode (1.2 m cable), electrode holder, 2 buffer sachets each for pH 4.01, 7, 9.21, 10.01, EasyDirect pH activation key, test certificate, pH measurement guide
Languages Chinese, Chinese (TW), Croatian, Czech, Danish, Dutch, English (GB), English (US), French, German, Hungarian, Indonesian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese (BR), Russian, Slovak, Slovenian, Spanish, Swedish, Thai, Turkish, Vietnamese
Memory 250 analyses (20.000 data points)
Model S500 Kit - Fluoride Measurement
Product Type Meter with Fluoride Kit
Range (Concentration) 0.001 to 999999mg/L ppm, 0.0001 to 100mol/L %, 0.001 to 100000mmol/L
Range (ISE) 0 to 999999mg/L
Resolution (Conductivity) 0.001 to 1
Resolution (mV) 0.1/1mV
Resolution (Resistivity) 0.001 to 1
Resolution (Salinity) 0.001 to 1
Resolution (TDS) 0.001 to 1
Temperature Accuracy ±0.1°C
Temperature Resolution 0.1°C