missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Mevalonolactone-4,4,5,5-d4, CDN
Click to view available options
Quantity:
0.025 g
0.05 g
Unit Size:
0.03g
0.05g
Specifications
Specifications
| Chemical Name or Material | (±)-Mevalonolactone-4,4,5,5-d4 |
| CAS | 349553-98-6 |
| Molecular Formula | C6 D4 H6 O3 |
| Quantity | 0.025 g |
| Synonym | 2 H-Pyran-2-one-5,6-d2, tetrahydro-5,6-d2-4-hydroxy-4-methyl- (9 CI), Tetrahydro-4-hydroxy-4-methyl-2 H-pyran-2-one-5,5,6,6-d4, (±)-Mevalolactone-d4, (±)-Mevalonolactone-d4, DL-Mevalonic acid lactone-d4, (±)-Mevalolactone-4,4,5,5-D4, (±)-Mevalolactone-4,4,5,5-D4, (±)-Mevalolactone-4,4,5,5-D4, (±)-Mevalolactone-4,4,5,5-D4 |
| InChI Formula | InChI=1 S/C6H10O3/c1-6(8)2-3-9-5(7)4-6/h8H,2-4H2,1H3/i2D2,3D2 |
| SMILES | [2 H]C1([2 H])OC(=O)CC(C)(O)C1([2 H])[2 H] |
| IUPAC Name | 5,5,6,6-tetradeuterio-4-hydroxy-4-methyloxan-2-one |
| Molecular Weight (g/mol) | 134.17 |
| Formula Weight | 134.0881 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction