Learn More
Invitrogen™ MFAP3L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553693
Description
Immunogen sequence: TNSTLNGTNV VLGSVPVIIA RTDHIIVKEG NSALINCSVY GIPDPQFKWY NSIGKLLKEE EDEKERGGGK WQMHDSGLLN ITKVSFSDRG KYTCVASNIY GTVNNTVTLR VIFTSGD Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 90%.
MFAP3L is a protein coding gene. May participate in the nuclear signaling of EGFR and MAPK1/ERK2. May a have a role in metastasis.
Specifications
| MFAP3L | |
| Polyclonal | |
| Unconjugated | |
| MFAP3L | |
| 4933428A15Rik; 5430405D20Rik; AI461995; AW125052; HSD39; HSD-39; KIAA0626; Mfap3l; microfi brillar-associated protein 3-like; microfibril associated protein 3 like; microfibrillar associated protein 3 like; microfibrillar-associated protein 3-like; mKIAA0626; NYD-sp9; Testis development protein NYD-SP9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9848 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75121 | |
| MFAP3L | |
| Recombinant protein corresponding to Human MFAP3L. Recombinant protein control fragment (Product #RP-91059). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.