missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFN2 Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 82673-2-RR-100UL
This item is not returnable.
View return policy
Description
The MFN2 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human MFN2. This antibody recognizes human, rat antigen. The MFN2 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
| MFN2 | |
| Recombinant Monoclonal | |
| 1000 μg/mL | |
| CMT2A; CMT2A2; CPRP1; HSG; KIAA0214; MARF | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| 86 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| 2A5 | |
| Unconjugated | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 9927 | |
| Store at -20°C. Stable for one year after shipment. | |
| Recombinant Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction