missing translation for 'onlineSavingsMsg'
Learn More

MgcRacGAP/RACGAP1 Antibody (JE41-19), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-32889

Product Code. 30114524

  • 496.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MgcRacGAP/RACGAP1 Monoclonal antibody specifically detects MgcRacGAP/RACGAP1 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MgcRacGAP/RACGAP1
Monoclonal
1 mg/mL
Western Blot 1:1000
GTPase activating protein, HsCYK-4, ID-GAP, KIAA1478, Male germ cell RacGap, MGCRACGAP, MgcRacGAPrac GTPase-activating protein 1, Rac GTPase activating protein 1
Synthetic peptide. (Uniprot: Q9H0H5)
100 μL
Core ESC Like Genes, Signal Transduction, Stem Cell Markers
29127
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot
JE41-19
Unconjugated
1*TBS (pH7.4), 0.05% BSA and 40% Glycerol
Rabbit
Protein A purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.