missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGP Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10734-1-AP-20UL
This item is not returnable.
View return policy
Description
MGP is associates with the organic matrix of bone and cartilage. MGP is thought to act as an inhibitor of bone formation.Specifications
| MGP | |
| Polyclonal | |
| Unconjugated | |
| MGP | |
| GIG36, matrix Gla protein, MGLAP, MGP, NTI | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4256 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P08493 | |
| MGP | |
| MGP Fusion Protein Ag1091 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction