missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MGST3 Antibody (SR2438), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27732
This item is not returnable.
View return policy
Description
MGST3 Monoclonal antibody specifically detects MGST3 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| MGST3 | |
| Monoclonal | |
| Unconjugated | |
| 50 mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 4259 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| SR2438 | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunoprecipitation 1:20 | |
| EC 2.5.1.18, GST-III, microsomal glutathione S-transferase 3, microsomal glutathione S-transferase III, Microsomal GST-3, Microsomal GST-III | |
| A synthetic peptide of human MGST3 (Uniprot #: O14880) | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction