missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class I Antibody (2G5), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65938AF750
This item is not returnable.
View return policy
Description
MHC Class I Monoclonal antibody specifically detects MHC Class I in Human, Mouse, Rat, Porcine, Bovine, Chinese Hamster, Guinea Pig, Hamster, Sheep samples. It is validated for Flow Cytometry
Specifications
| MHC Class I | |
| Monoclonal | |
| Alexa Fluor 750 | |
| DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1 | |
| Purified H-2Kb and H-2Db MHC-I molecules. | |
| 0.125 mL | |
| Adaptive Immunity, Asthma, Cell Biology, Core ESC Like Genes, Diabetes Research, DNA Repair, DNA replication Transcription Translation and Splicing, Immunology, Stem Cell Markers | |
| 3133 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry | |
| 2G5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine, Chinese Hamster, Guinea Pig, Hamster, Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction