missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MHC Class I (H-2Db) Monoclonal Antibody (28-14-8), NovaFluor™ Blue 690, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ M038T03B09-A

 View more versions of this product

Product Code. 30055002

  • 344.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This protein is an MHC class I alloantigen, which is involved in antigen presentation to T cells expressing CD3/TCR and CD8. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MHC Class I (H-2Db)
Monoclonal
0.1 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P01895, P01896, P01897, P01899, P01900, P14426, P14427
H2-D1
Affinity chromatography
RUO
14964
4°C, store in dark, DO NOT FREEZE!
Liquid
IgG2a κ
Flow Cytometry
28-14-8
NovaFluor Blue 690
H2-D1
H-2 class I histocompatibility antigen, D-B alpha chain; H-2D; H2-D; H-2D cell surface glycoprotein; H-2D(B); H2D1; H2-D1; H2DB; H2-DC1-beta transplantation antigen protein; H2L; H2LD; histocompatibility 2, D region locus 1; MHC class I antigen; MHC class I H2 antigen; MHC class I H-2Dp; MHC H2-D-q alpha-chain; Q5k protein
Mouse
100 μg
Primary
Mouse
Antibody
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.