missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC class II (I-A/I-E) Antibody (M5/114.15.2), Allophycocyanin, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-43312APC
This item is not returnable.
View return policy
Description
MHC class II (I-A/I-E) Monoclonal antibody specifically detects MHC class II (I-A/I-E) in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| MHC class II (I-A/I-E) | |
| Monoclonal | |
| APC | |
| E-alpha-f;H-2 class II histocompatibility antigen, E-K alpha chain;H-2 class II histocompatibility antigen, E-U alpha chain;H-2E;H2-Ea;H-2Ea;H2-Ea-ps;I;Ia;Ia3;Ia-3;I-E;I-Ealpha;MHC-H2-Ea | |
| The immunogen for this antibody was MHC Class II. | |
| 0.1 mL | |
| Adaptive Immunity, Asthma, B Cell Development and Differentiation Markers, Cell Biology, Diabetes Research, Immune System Diseases, Immunology, Transcription Factors and Regulators | |
| 100504404 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| M5/114.15.2 | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction