Learn More
MICA/B Antibody (159207), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB13001MFV450
This item is not returnable.
View return policy
Description
MICA/B Monoclonal antibody specifically detects MICA/B in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Blocking Assay, CyTOF
Specifications
MICA/B | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Flow Cytometry, Immunohistochemistry, Blocking Assay, CyTOF | |
159207 | |
Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
MHC class I chain-related protein A, MHC class I chain-related protein B, MIC, MICA, MICB, PERB11, PERB11.1, PERB11.2 | |
BaF3 mouse pro-B cell line transfected with human MICA, Accession # Q29983 | |
RUO | |
100507436 | |
Store at 4C in the dark. |