Learn More
Eppendorf™ Centrifuge 5430/5430 R
BIOPHARMAGREENER_CHOICE
Product Code. 15865073 Shop All Eppendorf Products
Change view
Click to view available options
Electrical Requirements:
230 V, 50/60 Hz (EU)
230 V/50-60 Hz (EU)
Includes:
With Rotor A-2-MTP
With Rotor FA-45-30-11 incl. rotor lid
Without rotor
Controller Type:
Keypad
Rotary Knob
Model:
Centrifuge 5430
Centrifuge 5430 (EU-IVD)
Centrifuge 5430 R
Centrifuge 5430 R (EU-IVD)
Unit Size:
1 set
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Electrical Requirements Includes Controller Type Model unitSize
15865073 230 V, 50/60 Hz (EU) With Rotor FA-45-30-11 incl. rotor lid Rotary Knob Centrifuge 5430 R 1 set
10326924 230 V, 50/60 Hz (EU) With Rotor FA-45-30-11 incl. rotor lid Rotary Knob Centrifuge 5430 (EU-IVD) Each
10091594 230 V, 50/60 Hz (EU) Without rotor Keypad Centrifuge 5430 (EU-IVD) Each
10329443 230 V/50-60 Hz (EU) Without rotor Rotary Knob Centrifuge 5430 R (EU-IVD) Each
15825073 230 V, 50/60 Hz (EU) With Rotor FA-45-30-11 incl. rotor lid Keypad Centrifuge 5430 R 1 set
15845073 230 V, 50/60 Hz (EU) Without rotor Keypad Centrifuge 5430 R Each
15885073 230 V, 50/60 Hz (EU) Without rotor Rotary Knob Centrifuge 5430 R Each
15865063 230 V, 50/60 Hz (EU) With Rotor FA-45-30-11 incl. rotor lid Keypad Centrifuge 5430 1 set
15845063 230 V, 50/60 Hz (EU) Without rotor Keypad Centrifuge 5430 Each
16692832 230 V, 50/60 Hz (EU) With Rotor A-2-MTP Rotary Knob Centrifuge 5430 Each
15885063 230 V, 50/60 Hz (EU) With Rotor FA-45-30-11 incl. rotor lid Rotary Knob Centrifuge 5430 1 set
15805073 230 V, 50/60 Hz (EU) Without rotor Rotary Knob Centrifuge 5430 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Regulatory: This product is under eco-contribution tax.
Product Code. 15865073 Supplier Eppendorf™ Supplier No. 5428000410
Equipment Monthly Promotion
Use promo code 28129
View all promotions
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
The unique, cross-over Eppendorf Centrifuge 5430 and 5430 R combine the small footprint of a microcentrifuge with the versatility of a multipurpose centrifuge. Extremely compact, they can accommodate rotors for microplates and 15/50 mL conical tubes as well as Eppendorf tubes and PCR strips.
  • Max. rotor capacity: 48 × 1.5/2.0 mL, 6 × 50 mL, 2 × MTP
  • Max. speed: 30,130 × g (17,500 rpm)
  • Remarkable versatility with 12 different rotors
  • Soft-touch one-finger lid closure for ergonomic operation
  • Menu-driven, multi lingual operation menu (English, German, French, Spanish) with large backlit display
  • 5 program keys for easy access to routine programs
  • Saves up to 50 user-defined programs
  • Automatic rotor recognition and imbalance detection for maximized operational safety
  • Eppendorf QuickLock™ – System for quickly opening and closing the rotor lid
  • TRUSTED_SUSTAINABILITY
    Specifications
    Capacity 30 x 2 mL
    Max. Speed 17,500 rpm
    Max. RCF 30,130 x g
    Refrigerated Yes
    Height (English) Exterior 11.7 in.
    Height (Metric) Exterior 29 cm
    Noise Level <54 dB(A) with Rotor F-45-30-11
    Safety Features Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, automatic rotor recognition, certified aerosol-tight rotor lids, automatic imbalance recognition
    Timer 30 sec. to 99:59 hrs., with continuous run function
    Width (English) Exterior 15 in.
    Width (Metric) Exterior 38 cm
    Wattage 1.050 W
    Certifications/Compliance UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers
    Includes With Rotor FA-45-30-11 incl. rotor lid
    Dimensions (L x W x H) Exterior 380 x 640 x 290 mm
    Electrical Requirements 230 V, 50/60 Hz (EU)
    Controller Type Rotary Knob
    Rotor Included Yes
    Standards EN ISO 12100, EN 50581, EN 61010-1, IEC 61010-2-020, EN 61326-1, EN 55011, EN 378-1, EN 378-2, 2 EN 50581 (refer to manual for more details)
    Frequency 50/60 Hz
    Voltage 230 V
    Model Centrifuge 5430 R
    Display Type Large, brightly lit LCD
    Temperature Range (Metric) -11°C to +40°C