missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- ASTM D6319, Category III, CE 0493 certified
- Comfortable and reliable protection for healthcare, laboratory workers, or any glove wearers seeking an ideal fit
- Lightweight nitrile formulation ensures enhanced comfort and tactile sensitivity, while textured fingertips provide better grip
- Packaged with 200 gloves/box
- For use in healthcare and chemotherapy applications
Specifications
Specifications
| Certifications/Compliance | ASTM D6319, Category III, CE 0493, ISO 13485, ISO 9001 |
| Certifications/Compliance (Europe) | EN 16523-1, EN 374-2 and -4, EN 420:2003 + A1:2009, EN 455 1-4, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-1:2016, EN ISO 374-5:2016, EN ISO 374-5:2016 |
| Accelerator Free | No |
| Applications | Healthcare |
| AQL | 1.5 |
| Color | Blue |
| Disposable | Single-use |
| Exterior Finish | Fingertip Textured |
| Finger Thickness (English) | 3.9 mil |
| Finger Thickness (Metric) | 0.1 mm |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?