Learn More
Hamilton Microlab 700, Dual Diluter Disposable Tip Hand Probe
Product Code. 31257772
Change view
Click to view available options
Product Type:
Convertible Tip Hand Probe
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
31257772 Convertible Tip Hand Probe Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 31257772 Supplier Hamilton Supplier No. 10103470
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Semi automatic high precision liquid handling system designed for accurate dispensing tasks. Ideal for applications where cross contamination must be avoided.
  • Microlab 700 Dual Syringe Diluter with Disposable Tip Hand Probe (PN: 10103470)
  • Designed to improve throughput and consistency in manual pipetting tasks
  • Enables semi-automated liquid handling, dilution, and dispensing
  • Delivers over 99% accuracy for precise results
  • Features a highly inert fluid path suitable for sensitive applications
Specifications
Product Type Convertible Tip Hand Probe
For Use With (Equipment) Microlab 700 Series