Learn More
Micronova™ Scoops
Product Code. 13177656 Shop All Micronova Products
Change view
Click to view available options
Capacity (Metric):
75 mL
1100 mL
Unit Size:
Pack of 36
Pack of 47
Pack of 6
Autoclavable:
Autoclavable
Not Autoclavable
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize Autoclavable
13177656 75 mL Pack of 47 -
13137656 1100 mL Pack of 6 Autoclavable
13157656 - Pack of 36 Autoclavable
13147656 1100 mL Pack of 6 Not Autoclavable
13167656 - Pack of 36 Not Autoclavable
13187656 75 mL Pack of 47 Not Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 missing translation for 'options'
Product Code. 13177656 missing translation for 'mfr' Micronova™ missing translation for 'supplierNo' SCP07
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Durable polypropylene scoop, available in sizes from 75 cc to 1100 cc. Ideal for scooping and sampling powders, granules, and pastes.
Micronova durable polypropylene scoops are available in sizes from 75 cc to 1100 cc and are ideal for scooping and sampling powders, granules, and pastes. Irradiated option available for sterile environments.
  • Individually bagged
  • Irradiated option available
Specifications
Product Type Scoop
Capacity (Metric) 75 mL
Material Polypropylene
Capacity (English) 2.53 oz.