Learn More
Sartorius Stedim Biotech™ Microsart™ Cellulose Nitrate Membrane Filter Units, Gridded, Grey/White, 0.45μm
Sartorius Stedim Biotech™ Microsart™ Cellulose Nitrate Membrane Filter Units, Gridded, Black/White 0.45μm combine a funnel, a filter base and a gridded membrane filter in one unit.
Brand: Sartorius Stedim Biotech™ 16D03--10-H6-ACG
Additional Details : Weight : 1.25000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II
Description
- Ready-to-use, sterile filter units combine a funnel, a filter base, and a gridded membrane filter in one unit.
- Suitable for the detection and enumeration of microorganisms in pharmaceuticals, cosmetics, food, beverages, water, and other liquids.
- Suitable for use in pharma/biotech, food and beverage, environmental/water testing.
- Suitable for microbial enumeration.
- Sterilized by gamma irradiation.
Specifications
Gray/White Grid | |
Polypropylene Funnel, ABS Base | |
0.45 μm | |
27 individually packed |
Funnel capacity: 100mL | |
Cellulose Nitrate | |
Sterile | |
47 mm |