missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Microsart™ Cellulose Nitrate Membrane Filter Units, Gridded, Grey/White, 0.45μm

Sartorius Stedim Biotech™ Microsart™ Cellulose Nitrate Membrane Filter Units, Gridded, Black/White 0.45μm combine a funnel, a filter base and a gridded membrane filter in one unit.

Brand:  Sartorius Stedim Biotech™ 16D03--25-H6--BS

Additional Details : Weight : 0.80000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II

 View more versions of this product

Product Code. 15922570

  • 1490.00€ / Pack of 16
Estimated Shipment: 15-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Ready-to-use, sterile filter units combine a funnel, a filter base, and a gridded membrane filter in one unit.
  • Suitable for the detection and enumeration of microorganisms in pharmaceuticals, cosmetics, food, beverages, water, and other liquids.
  • Suitable for use in pharma/biotech, food and beverage, environmental/water testing.
  • Suitable for microbial enumeration.
  • Sterilized by gamma irradiation.
Specifications

Specifications

Gray/White Grid
Polypropylene Funnel, ABS Base
0.45 μm
6 bags of 8
Funnel capacity: 250mL
Cellulose Nitrate
Sterile
47 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions