Learn More
Kern Sohn Microscope Camera ODC
Product Code. 17309541
Encompass_Preferred
Change view
Click to view available options
Port Type:
USB 2.0
USB 3.0
Resolution:
20 MP
5.1 MP
Size:
1 in. Sensor
1/2.5 in. Sensor
Compact, 1 in. Sensor
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Port Type Resolution Size unitSize
17309541 USB 2.0 5.1 MP 1/2.5 in. Sensor Each
17319541 USB 3.0 5.1 MP 1/2.5 in. Sensor Each
17329541 USB 2.0 20 MP 1 in. Sensor Each
17339541 USB 2.0 20 MP Compact, 1 in. Sensor Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Regulatory: This product is under eco-contribution tax.
Product Code. 17309541 Supplier Kern Sohn Supplier No. ODC 825
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
Specialists in microscopy for measurement, counting, documentation, archiving and image processing.
  • The universal microscope cameras can be used anywhere and can be connected to the microscope as well as to a laptop or PC using the USB cable (USB 2.0)
  • Through the proven CMOS technology, in connection with the USB 2.0 the images are shown quickly and clearly
  • These cameras are also ideal for more demanding applications, such as, for example, darkfield, phase contrast and for fluorescence applications.
  • Your daily work is made significantly easier with the very best synchronisation, a high frame rate as well as stable image performance together with our Microscope VIS software which we deliver with the product.
  • The power supply is through the USB cable, which means that no additional power supply is required
  • As well as the camera, the delivery includes our multi-lingual KERN Microscope VIS Basic KERN OXM 901, a USB cable (Length: 2000 mm) various eyepiece adapters and an object micrometer to calibrate the software.
  • Please order the appropriate C-mount adapter to fit your KERN microscope now.
Specifications
For Use With (Equipment) Microscopes
Port Type USB 2.0
Resolution 5.1 MP
Sensor Types CMOS
Size 1/2.5 in. Sensor