Learn More
Cytiva Microspin Column
Product Code. 11783299 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 Pack
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11783299 100 Pack Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11783299 Supplier Cytiva Supplier No. 27356501
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For the rapid purification of DNA for use in a wide range of applications
  • Empty microspin columns with screw caps for use with chromatography media of choice such as Sephadex™ for desalting, DNA purification etc
  • Empty microspin columns provide flexibilty with media choice and bed volume allowing process optimization
TRUSTED_SUSTAINABILITY
Specifications
Format Spin Column
Isolation Technology Spin Column
For Use With (Application) Rapid purification of DNA, desalting/buffer exchange of DNA
Product Type Column
Quantity 100 Pack