Learn More
Scientific Industries SI™ Microtube Foam Insert
Product Code. 15577325 Shop All Ohaus Products
Change view
Click to view available options
No. of Positions:
60
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Positions Unit Size
15577325 60 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15577325 Supplier Scientific Industries SI™ Supplier No. 83041418
  • ONLINE EXCLUSIVE PRICE
    68.53€
    49.84€ / Pack of 1
    Save 18.69€
    27% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Holds vessels in place while mixing. Scientific Industries SI™ Microtube Foam Insert fits snugly inside the Vortex-Genie 2 and Vortex-Genie Pulse 152mm platform attachment.
  • Accommodates up to 60 microtubes.
  • Fits into 152mm Platform attachment
  • Included in the Multiple Sample Starter Set
  • For use with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse
TRUSTED_SUSTAINABILITY
Specifications
Material High-density foam
For Use With (Equipment) Vortex-Genie 2 family of mixers, Vortex-Genie Pulse
No. of Positions 60
Requires Platform Attachment 15577315