Learn More
Nickel Electro™ Mild Steel Tongs Bow
Product Code. 11582692 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Length (Metric):
400 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11582692 400 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11582692
  • ONLINE EXCLUSIVE PRICE
    13.15€
    10.97€ / Pack of 1
    Save 2.18€
    16% Off
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Laboratory mild steel bow tong with bright finish.
  • Laboratory bow tong
  • Mild steel
  • 400 mm
  • With bright finish
Specifications
Product Type Laboratory Bow Tongs
Sterility Non-sterile
Length (Metric) 400 mm
Material Mild Steel