Learn More
Nickel Electro™ Mild Steel Tongs Straight
Product Code. 11572692 Shop All Fisherbrand Products
Change view
Click to view available options
Length (Metric):
400 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
11572692 400 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11572692
  • ONLINE EXCLUSIVE PRICE
    13.60€
    11.41€ / Pack of 1
    Save 2.19€
    16% Off
In Stock
Add to basket
Add to basket
Laboratory straight tong with bright finish.
  • Laboratory straight tong
  • Mild steel
  • 381 mm
  • With bright finish
Specifications
Sterility Non-sterile
Length (Metric) 400 mm
Material Mild Steel
Type General Purpose Tongs