Description
The Millicell™ disposable hemocytometer is used to manually count cells. It consists of a surface-patterned two or four enclosed chambers with ports for sample injection. Count cells in the same way as you do with the Neubauer Improved counting chamber.
The Neubauer Improved counting chamber consists of 9 large squares, each measuring 1 x 1 mm. Each square has a total volume of 0.1 mm3. The cells are distributed over 3 x 3 large squares, each with 1 mm edge length and with a surface area of 1 mm2.
With the Millicell™ disposable hemocytometer, you inject 10 μL of sample, stained or unstained, into the desired chamber.
Tekniske data
Tekniske data
| Product Type | Hemacytometer |
| Grid Type | Neubauer Improved |
| For Use With (Application) | Cell counting |
| Includes | 2 Chambers/Slide |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.