missing translation for 'onlineSavingsMsg'
Learn More

Milrinone Lactate, TRC

Product Code. 30479331
Change view
Click to view available options
Quantity:
25 mg
50 mg
200 mg
Unit Size:
200mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30479331 25 mg 25mg
30465428 200 mg 200mg
30478028 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30479331 Supplier Toronto Research Chemicals Supplier No. TRCM34468325MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100286-97-3
Molecular Formula C12H9N3O . x(C3H6O3)
Molecular Weight (g/mol) 301.3
IUPAC Name 2-hydroxypropanoic acid;6-methyl-2-oxo-5-pyridin-4-yl-1H-pyridine-3-carbonitrile
SMILES CC(O)C(=O)O.CC1=C(C=C(C#N)C(=O)N1)c2ccncc2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H9N3O.C3H6O3/c1-8-11(9-2-4-14-5-3-9)6-10(7-13)12(16)15-8;1-2(4)3(5)6/h2-6H,1H3,(H,15,16);2,4H,1H3,(H,5,6)
Formula Weight 301.1063
Chemical Name or Material Milrinone Lactate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.